Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
Chromosome Location
Chr.6: 44158811 - 44184402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007058.3 NP_008989.2 21 - 22 2171 102
XM_006714984.1 XP_006715047.1 21 - 22 2174 102
XM_011514274.1 XP_011512576.1 21 - 22 2255 102
XM_011514275.1 XP_011512577.1 22 - 23 2297 102
GenBank mRNA AJ242832.1 - 20 - 21 2125 102
AK314054.1 - 21 - 22 2151 102
BC033733.1 - 21 - 22 2142 102

View Full Product Details