The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 15 - 16 | 2188 | 89 |
XM_011539637.2 | XP_011537939.1 | 15 - 16 | 3374 | 89 | |
XM_011539638.2 | XP_011537940.1 | 15 - 16 | 1961 | 89 | |
XM_017016072.1 | XP_016871561.1 | 15 - 16 | 1959 | 89 | |
GenBank mRNA | AB033100.2 | - | 15 - 16 | 2186 | 89 |
BC015004.1 | - | 1 - 2 | 48 | 89 | |
BC040163.1 | - | 13 - 14 | 1691 | 89 | |
BC048195.1 | - | 1 - 2 | 37 | 89 | |
BC136375.1 | - | 15 - 16 | 2070 | 89 | |
BC136376.1 | - | 15 - 16 | 2070 | 89 |