Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
Chromosome Location
Chr.15: 65578757 - 65611289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136043.2 NP_001129515.1 3 - 4 439 108
NM_001207058.1 NP_001193987.1 3 - 4 418 108
NM_001207059.1 NP_001193988.1 2 - 3 298 108
NR_045104.1 3 - 4 582 108
NR_045105.2 3 - 4 836 108
NR_045106.2 3 - 4 743 108
XM_006720708.3 XP_006720771.1 3 - 4 518 108
XM_006720709.3 XP_006720772.1 3 - 4 583 108
XM_006720711.2 XP_006720774.1 2 - 3 442 108
XM_011522101.2 XP_011520403.1 3 - 4 264 108
XM_017022644.1 XP_016878133.1 3 - 4 663 108
GenBank mRNA AK027475.1 - 3 - 4 424 108
AK293207.1 - 2 - 3 251 108
AK296134.1 - 3 - 4 809 108
AK301033.1 - 3 - 4 281 108
AK301742.1 - 3 - 4 260 108
AK310857.1 - 3 - 4 428 108
BC007991.2 - 3 - 4 409 108
DC298728.1 - 2 - 3 251 108
DC384988.1 - 3 - 4 335 108

View Full Product Details