The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136043.2 | NP_001129515.1 | 6 - 7 | 859 | 131 |
NM_001207058.1 | NP_001193987.1 | 6 - 7 | 838 | 131 | |
NM_001207059.1 | NP_001193988.1 | 5 - 6 | 718 | 131 | |
NR_045104.1 | 6 - 7 | 1002 | 131 | ||
NR_045105.2 | 6 - 7 | 1256 | 131 | ||
NR_045106.2 | 6 - 7 | 1163 | 131 | ||
NR_045901.1 | 5 - 6 | 894 | 131 | ||
XM_006720708.3 | XP_006720771.1 | 6 - 7 | 938 | 131 | |
XM_006720709.3 | XP_006720772.1 | 6 - 7 | 1003 | 131 | |
XM_006720711.2 | XP_006720774.1 | 5 - 6 | 862 | 131 | |
XM_011522101.2 | XP_011520403.1 | 6 - 7 | 684 | 131 | |
XM_017022644.1 | XP_016878133.1 | 6 - 7 | 1083 | 131 | |
GenBank mRNA | AK027475.1 | - | 6 - 7 | 844 | 131 |
AK293207.1 | - | 5 - 6 | 671 | 131 | |
AK296134.1 | - | 6 - 7 | 1229 | 131 | |
AK301033.1 | - | 6 - 7 | 701 | 131 | |
AK301222.1 | - | 5 - 6 | 847 | 131 | |
AK301742.1 | - | 6 - 7 | 680 | 131 | |
AL136662.1 | - | 6 - 7 | 1084 | 131 | |
BC007991.2 | - | 6 - 7 | 829 | 131 |