Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK1
Entrez Gene ID
65125
Gene Name
WNK lysine deficient protein kinase 1
Gene Aliases
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location
Chr.12: 752552 - 911452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184985.1 NP_001171914.1 21 - 22 6871 137
NM_014823.2 NP_055638.2 19 - 20 5350 137
NM_018979.3 NP_061852.3 21 - 22 6091 137
NM_213655.4 NP_998820.3 21 - 22 6847 137
XM_006719003.2 XP_006719066.1 21 - 22 6413 137
XM_011520997.2 XP_011519299.1 22 - 23 7701 137
XM_011520998.2 XP_011519300.1 22 - 23 7652 137
XM_011520999.2 XP_011519301.1 22 - 23 7655 137
XM_011521000.2 XP_011519302.1 22 - 23 7655 137
XM_011521001.2 XP_011519303.1 21 - 22 7376 137
XM_011521002.2 XP_011519304.1 21 - 22 7193 137
XM_011521003.2 XP_011519305.1 20 - 21 6917 137
XM_011521004.2 XP_011519306.1 20 - 21 6914 137
XM_011521005.2 XP_011519307.1 19 - 20 6139 137
XM_011521006.2 XP_011519308.1 20 - 21 6332 137
XM_011521007.2 XP_011519309.1 20 - 21 6329 137
XM_011521008.2 XP_011519310.1 18 - 19 5594 137
XM_011521009.2 XP_011519311.1 18 - 19 5591 137
XM_017019834.1 XP_016875323.1 20 - 21 5954 137
XM_017019835.1 XP_016875324.1 19 - 20 5873 137
XM_017019836.1 XP_016875325.1 19 - 20 5870 137
XM_017019837.1 XP_016875326.1 19 - 20 5678 137
XM_017019838.1 XP_016875327.1 19 - 20 5675 137
XM_017019839.1 XP_016875328.1 18 - 19 5594 137
GenBank mRNA AB002342.2 - 19 - 20 4505 137
AJ296290.1 - 21 - 22 5448 137
BC141881.1 - 13 - 14 2915 137
JQ358908.1 - 22 - 23 6687 137

View Full Product Details