The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320556.1 | NP_001307485.1 | 1 - 2 | 122 | 63 |
NM_001320557.1 | NP_001307486.1 | 1 - 2 | 122 | 63 | |
NM_001320558.1 | NP_001307487.1 | 1 - 2 | 122 | 63 | |
NM_025231.2 | NP_079507.1 | 1 - 2 | 122 | 63 | |
GenBank mRNA | AK025844.1 | - | 1 - 2 | 69 | 63 |
AK294017.1 | - | 1 - 2 | 69 | 63 | |
AK309837.1 | - | 1 - 2 | 69 | 63 | |
BC004255.1 | - | 1 - 2 | 43 | 63 | |
CB158915.1 | - | 1 - 2 | 51 | 63 | |
DB158261.1 | - | 1 - 2 | 122 | 63 |