The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321837.1 | NP_001308766.1 | 7 - 8 | 1011 | 101 |
NM_001321839.1 | NP_001308768.1 | 6 - 7 | 915 | 101 | |
NM_004056.5 | NP_004047.3 | 7 - 8 | 1011 | 101 | |
XM_017013818.1 | XP_016869307.1 | 8 - 9 | 1076 | 101 | |
GenBank mRNA | AK090655.1 | - | 7 - 8 | 983 | 101 |
AK289470.1 | - | 7 - 8 | 983 | 101 | |
AK293999.1 | - | 6 - 7 | 602 | 101 | |
AK314538.1 | - | 7 - 8 | 984 | 101 | |
AY075022.1 | - | 7 - 8 | 988 | 101 | |
BC069744.1 | - | 7 - 8 | 980 | 101 | |
BC069794.1 | - | 7 - 8 | 1015 | 101 | |
BC108929.2 | - | 7 - 8 | 936 | 101 | |
L04656.1 | - | 7 - 8 | 733 | 101 |