The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042410.1 | NP_001035869.1 | 12 - 13 | 2078 | 116 |
NM_001282792.1 | NP_001269721.1 | 9 - 10 | 1757 | 116 | |
NM_018089.2 | NP_060559.2 | 12 - 13 | 2151 | 116 | |
XM_005246663.2 | XP_005246720.1 | 12 - 13 | 2070 | 116 | |
XM_011511392.2 | XP_011509694.1 | 12 - 13 | 2148 | 116 | |
XM_017004420.1 | XP_016859909.1 | 12 - 13 | 2067 | 116 | |
GenBank mRNA | AA482007.1 | - | 2 - 3 | 116 | |
AF364318.1 | - | 11 - 12 | 2541 | 116 | |
AK001277.1 | - | 12 - 13 | 2087 | 116 | |
AK023206.1 | - | 12 - 13 | 2078 | 116 | |
AK056060.1 | - | 1 - 2 | 1585 | 116 | |
AK095304.1 | - | 9 - 10 | 2835 | 116 | |
AK225275.1 | - | 12 - 13 | 2076 | 116 | |
AK225309.1 | - | 12 - 13 | 2052 | 116 | |
BC000238.1 | - | 12 - 13 | 2033 | 116 | |
BC008948.2 | - | 12 - 13 | 2062 | 116 | |
BC028334.1 | - | 9 - 10 | 1679 | 116 | |
DQ412564.1 | - | 12 - 13 | 2087 | 116 |