Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD12
Entrez Gene ID
26090
Gene Name
abhydrolase domain containing 12
Gene Aliases
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2
Chromosome Location
Chr.20: 25294743 - 25390982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042472.2 NP_001035937.1 5 - 6 853 79
NM_015600.4 NP_056415.1 5 - 6 853 79
XM_005260698.1 XP_005260755.1 5 - 6 584 79
XM_011529214.2 XP_011527516.1 5 - 6 584 79
XM_011529215.1 XP_011527517.1 6 - 7 704 79
XM_017027796.1 XP_016883285.1 6 - 7 704 79
GenBank mRNA AK075023.1 - 5 - 6 584 79
AK290815.1 - 5 - 6 584 79
AK293495.1 - 6 - 7 470 79
AL117442.1 - 2 - 3 121 79
BC014049.2 - 5 - 6 698 79

View Full Product Details