The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007058.3 | NP_008989.2 | 14 - 15 | 1680 | 81 |
XM_006714984.1 | XP_006715047.1 | 14 - 15 | 1683 | 81 | |
XM_006714985.1 | XP_006715048.1 | 14 - 15 | 1683 | 81 | |
XM_006714987.1 | XP_006715050.1 | 14 - 15 | 1683 | 81 | |
XM_011514274.1 | XP_011512576.1 | 14 - 15 | 1764 | 81 | |
XM_011514275.1 | XP_011512577.1 | 15 - 16 | 1806 | 81 | |
GenBank mRNA | AJ242832.1 | - | 13 - 14 | 1634 | 81 |
AK300107.1 | - | 5 - 6 | 606 | 81 | |
AK309883.1 | - | 4 - 5 | 992 | 81 | |
AK314054.1 | - | 14 - 15 | 1660 | 81 | |
BC033733.1 | - | 14 - 15 | 1651 | 81 |