The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 10 - 11 | 1919 | 83 |
NM_001321454.1 | NP_001308383.1 | 11 - 12 | 2030 | 83 | |
NM_001321455.1 | NP_001308384.1 | 11 - 12 | 2076 | 83 | |
NM_001321458.1 | NP_001308387.1 | 11 - 12 | 2046 | 83 | |
NM_001321459.1 | NP_001308388.1 | 11 - 12 | 2122 | 83 | |
NM_001321460.1 | NP_001308389.1 | 11 - 12 | 2119 | 83 | |
NM_016316.3 | NP_057400.1 | 10 - 11 | 1922 | 83 | |
NR_135649.1 | 11 - 12 | 2119 | 83 | ||
NR_135650.1 | 11 - 12 | 2076 | 83 | ||
NR_135651.1 | 10 - 11 | 1919 | 83 | ||
NR_135652.1 | 10 - 11 | 1922 | 83 | ||
NR_135653.1 | 10 - 11 | 1922 | 83 | ||
XM_011511341.2 | XP_011509643.1 | 4 - 5 | 539 | 83 | |
XM_017004309.1 | XP_016859798.1 | 11 - 12 | 2443 | 83 | |
XM_017004310.1 | XP_016859799.1 | 11 - 12 | 1809 | 83 | |
XM_017004311.1 | XP_016859800.1 | 11 - 12 | 1809 | 83 | |
XM_017004312.1 | XP_016859801.1 | 11 - 12 | 2022 | 83 | |
XM_017004313.1 | XP_016859802.1 | 10 - 11 | 3648 | 83 | |
XM_017004314.1 | XP_016859803.1 | 7 - 8 | 1299 | 83 | |
XM_017004315.1 | XP_016859804.1 | 7 - 8 | 1283 | 83 | |
GenBank mRNA | AB047646.1 | - | 10 - 11 | 1844 | 83 |
AF151538.1 | - | 10 - 11 | 1875 | 83 | |
AF206019.1 | - | 10 - 11 | 1884 | 83 | |
AF357886.1 | - | 10 - 11 | 1840 | 83 | |
AJ131720.1 | - | 2 - 3 | 241 | 83 | |
AK025176.1 | - | 2 - 3 | 201 | 83 | |
AK307719.1 | - | 10 - 11 | 1833 | 83 | |
BC037734.1 | - | 11 - 12 | 2057 | 83 | |
BC130411.1 | - | 9 - 10 | 1690 | 83 | |
BP332286.1 | - | 4 - 5 | 386 | 83 |