Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
Chromosome Location
Chr.22: 50274979 - 50307637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012401.3 NP_036533.2 32 - 33 5292 56
XM_005261909.1 XP_005261966.1 33 - 34 5340 56
XM_005261910.2 XP_005261967.1 33 - 34 5351 56
XM_005261911.4 XP_005261968.1 32 - 33 5290 56
XM_006724413.2 XP_006724476.1 34 - 35 5410 56
XM_011530682.2 XP_011528984.1 33 - 34 5491 56
XM_017028703.1 XP_016884192.1 34 - 35 5391 56
XM_017028704.1 XP_016884193.1 33 - 34 5344 56
GenBank mRNA AB002313.1 - 31 - 32 5209 56
AK025415.1 - 7 - 8 879 56
AK025701.1 - 19 - 20 2660 56
AK056543.1 - 15 - 16 2461 56
AK074932.1 - 14 - 15 1928 56
AK123131.1 - 4 - 5 873 56
AK126394.1 - 13 - 14 2001 56
BC004542.2 - 9 - 10 1111 56
BC110366.1 - 4 - 5 529 56
BT006887.1 - 9 - 10 1095 56

View Full Product Details