The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012401.3 | NP_036533.2 | 8 - 9 | 1972 | 67 |
XM_005261909.1 | XP_005261966.1 | 9 - 10 | 2020 | 67 | |
XM_005261910.2 | XP_005261967.1 | 9 - 10 | 2031 | 67 | |
XM_005261911.4 | XP_005261968.1 | 8 - 9 | 1970 | 67 | |
XM_006724413.2 | XP_006724476.1 | 10 - 11 | 2090 | 67 | |
XM_011530682.2 | XP_011528984.1 | 9 - 10 | 2171 | 67 | |
XM_017028703.1 | XP_016884192.1 | 10 - 11 | 2071 | 67 | |
XM_017028704.1 | XP_016884193.1 | 9 - 10 | 2024 | 67 | |
GenBank mRNA | AB002313.1 | - | 7 - 8 | 1889 | 67 |