The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278596.1 | NP_001265525.1 | 20 - 21 | 2047 | 81 |
NM_001278597.1 | NP_001265526.1 | 20 - 21 | 2065 | 81 | |
XM_006718988.3 | XP_006719051.1 | 18 - 19 | 2322 | 81 | |
XM_011520975.2 | XP_011519277.1 | 18 - 19 | 4608 | 81 | |
XM_011520978.2 | XP_011519280.1 | 17 - 18 | 1920 | 81 | |
XM_017019672.1 | XP_016875161.1 | 18 - 19 | 2322 | 81 | |
GenBank mRNA | AJ272204.1 | - | 20 - 21 | 2065 | 81 |
AK024010.1 | - | 3 - 4 | 329 | 81 | |
FJ384095.1 | - | 20 - 21 | 2021 | 81 |