The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278596.1 | NP_001265525.1 | 23 - 24 | 2444 | 69 |
NM_001278597.1 | NP_001265526.1 | 23 - 24 | 2462 | 69 | |
XM_006718988.3 | XP_006719051.1 | 21 - 22 | 2719 | 69 | |
XM_011520975.2 | XP_011519277.1 | 21 - 22 | 5005 | 69 | |
XM_011520978.2 | XP_011519280.1 | 20 - 21 | 2317 | 69 | |
XM_017019672.1 | XP_016875161.1 | 21 - 22 | 2719 | 69 | |
GenBank mRNA | AJ272204.1 | - | 23 - 24 | 2462 | 69 |
AK024010.1 | - | 6 - 7 | 726 | 69 | |
FJ384095.1 | - | 23 - 24 | 2418 | 69 |