The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017545.2 | NP_060015.1 | 4 - 5 | 739 | 95 |
GenBank mRNA | AB024079.1 | - | 4 - 5 | 715 | 95 |
AF231916.1 | - | 4 - 5 | 739 | 95 | |
AF244134.1 | - | 4 - 5 | 738 | 95 | |
AK289423.1 | - | 4 - 5 | 784 | 95 | |
AK308963.1 | - | 4 - 5 | 751 | 95 | |
AK313839.1 | - | 4 - 5 | 757 | 95 | |
AL121739.1 | - | 4 - 5 | 767 | 95 | |
BC113665.1 | - | 4 - 5 | 752 | 95 | |
BC113667.1 | - | 4 - 5 | 752 | 95 |