The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005445.3 | NP_005436.1 | 1 - 2 | 145 | 117 |
GenBank mRNA | AF020043.1 | - | 1 - 2 | 110 | 117 |
AK289771.1 | - | 1 - 2 | 141 | 117 | |
AK315845.1 | - | 1 - 2 | 144 | 117 | |
BC047324.1 | - | 1 - 2 | 40 | 117 | |
CA439554.1 | - | 1 - 2 | 159 | 117 |