Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
Chromosome Location
Chr.5: 119071002 - 119250170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290321.2 NP_001277250.1 23 - 24 5872 137
NM_001290322.2 NP_001277251.1 23 - 24 7180 137
NM_005509.5 NP_005500.4 23 - 24 5872 137
XM_005271909.4 XP_005271966.1 24 - 25 5570 137
XM_005271910.4 XP_005271967.1 23 - 24 5915 137
XM_005271912.2 XP_005271969.1 24 - 25 5554 137
XM_011543212.2 XP_011541514.1 23 - 24 5915 137
XM_011543213.2 XP_011541515.1 24 - 25 5500 137
XM_011543214.2 XP_011541516.1 20 - 21 5081 137
XM_011543215.2 XP_011541517.1 19 - 20 4917 137
XM_017009141.1 XP_016864630.1 24 - 25 5570 137
XM_017009142.1 XP_016864631.1 24 - 25 5500 137
XM_017009143.1 XP_016864632.1 25 - 26 5749 137
XM_017009144.1 XP_016864633.1 23 - 24 5452 137
XM_017009145.1 XP_016864634.1 23 - 24 5322 137
XM_017009146.1 XP_016864635.1 23 - 24 6764 137
XM_017009147.1 XP_016864636.1 23 - 24 5915 137
XM_017009148.1 XP_016864637.1 22 - 23 5718 137
GenBank mRNA AB593142.1 - 23 - 24 7183 137
AJ005821.1 - 23 - 24 5469 137
BC144537.1 - 23 - 24 5415 137
BC150616.1 - 23 - 24 5415 137

View Full Product Details