Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DMXL1
- Entrez Gene ID
-
1657
- Gene Name
- Dmx like 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 119071002 - 119250170 on Build GRCh38
View Full Product Details