Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
Chromosome Location
Chr.5: 119071002 - 119250170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290321.2 NP_001277250.1 25 - 26 7166 101
NM_001290322.2 NP_001277251.1 25 - 26 8474 101
NM_005509.5 NP_005500.4 25 - 26 7166 101
XM_005271909.4 XP_005271966.1 26 - 27 6864 101
XM_005271910.4 XP_005271967.1 25 - 26 7209 101
XM_005271912.2 XP_005271969.1 26 - 27 6848 101
XM_011543212.2 XP_011541514.1 25 - 26 7209 101
XM_011543213.2 XP_011541515.1 26 - 27 6794 101
XM_011543214.2 XP_011541516.1 22 - 23 6375 101
XM_011543215.2 XP_011541517.1 21 - 22 6211 101
XM_017009141.1 XP_016864630.1 26 - 27 6864 101
XM_017009142.1 XP_016864631.1 26 - 27 6794 101
XM_017009143.1 XP_016864632.1 27 - 28 7043 101
XM_017009144.1 XP_016864633.1 25 - 26 6746 101
XM_017009145.1 XP_016864634.1 25 - 26 6616 101
XM_017009146.1 XP_016864635.1 25 - 26 8058 101
XM_017009147.1 XP_016864636.1 25 - 26 7209 101
XM_017009148.1 XP_016864637.1 24 - 25 7012 101
GenBank mRNA AB593142.1 - 25 - 26 8477 101
AJ005821.1 - 25 - 26 6763 101
BC144537.1 - 25 - 26 6709 101
BC150616.1 - 25 - 26 6709 101

View Full Product Details