Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
Chromosome Location
Chr.5: 119071002 - 119250170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290321.2 NP_001277250.1 27 - 28 7372 119
NM_001290322.2 NP_001277251.1 27 - 28 8680 119
NM_005509.5 NP_005500.4 27 - 28 7372 119
XM_005271909.4 XP_005271966.1 28 - 29 7070 119
XM_005271910.4 XP_005271967.1 27 - 28 7415 119
XM_005271912.2 XP_005271969.1 28 - 29 7054 119
XM_011543212.2 XP_011541514.1 27 - 28 7415 119
XM_011543213.2 XP_011541515.1 28 - 29 7000 119
XM_011543214.2 XP_011541516.1 24 - 25 6581 119
XM_011543215.2 XP_011541517.1 23 - 24 6417 119
XM_017009141.1 XP_016864630.1 28 - 29 7070 119
XM_017009142.1 XP_016864631.1 28 - 29 7000 119
XM_017009143.1 XP_016864632.1 29 - 30 7249 119
XM_017009144.1 XP_016864633.1 27 - 28 6952 119
XM_017009145.1 XP_016864634.1 27 - 28 6822 119
XM_017009146.1 XP_016864635.1 27 - 28 8264 119
XM_017009147.1 XP_016864636.1 27 - 28 7415 119
XM_017009148.1 XP_016864637.1 26 - 27 7218 119
GenBank mRNA AB593142.1 - 27 - 28 8683 119
AJ005821.1 - 27 - 28 6969 119
BC144537.1 - 27 - 28 6915 119
BC150616.1 - 27 - 28 6915 119

View Full Product Details