Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
Chromosome Location
Chr.1: 27872531 - 27886685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK094833.1 - 1 - 1 623 66

View Full Product Details