Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD12
Entrez Gene ID
26090
Gene Name
abhydrolase domain containing 12
Gene Aliases
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2
Chromosome Location
Chr.20: 25294743 - 25390982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042472.2 NP_001035937.1 9 - 10 1142 106
NM_015600.4 NP_056415.1 9 - 10 1142 106
XM_005260698.1 XP_005260755.1 9 - 10 873 106
XM_011529214.2 XP_011527516.1 9 - 10 873 106
XM_011529215.1 XP_011527517.1 10 - 11 993 106
XM_011529218.1 XP_011527520.1 7 - 8 552 106
XM_017027796.1 XP_016883285.1 10 - 11 993 106
XM_017027798.1 XP_016883287.1 8 - 9 672 106
GenBank mRNA AK075023.1 - 9 - 10 873 106
AK290815.1 - 9 - 10 873 106
AK293495.1 - 10 - 11 759 106
AL117442.1 - 6 - 7 410 106
BC014049.2 - 9 - 10 987 106

View Full Product Details