Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR13
Entrez Gene ID
64743
Gene Name
WD repeat domain 13
Gene Aliases
MG21
Chromosome Location
Chr.X: 48597492 - 48605194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166426.1 NP_001159898.1 7 - 8 2117 77
NM_017883.4 NP_060353.2 8 - 9 1782 77
NR_029427.1 8 - 9 1748 77
XM_006724539.1 XP_006724602.1 9 - 10 1435 77
XM_006724541.3 XP_006724604.1 9 - 10 1401 77
XM_011543947.2 XP_011542249.1 9 - 10 1395 77
GenBank mRNA AF158978.1 - 7 - 8 76
AF329819.4 - 8 - 9 1782 77
AK000570.1 - 3 - 4 428 77
AK225542.1 - 3 - 4 428 77
AK314739.1 - 8 - 9 1745 77
BC002507.2 - 5 - 6 844 77
BC067094.1 - 8 - 9 1429 77
BC080579.1 - 8 - 9 1511 77
BX649101.1 - 6 - 7 727 77
DQ907252.1 - 9 - 10 1474 77
DQ907253.1 - 7 - 8 1789 77
DQ907254.1 - 7 - 8 1783 77
DQ907255.1 - 7 - 8 1732 77

View Full Product Details