The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002517.3 | NP_002508.2 | 4 - 5 | 623 | 132 |
XM_006723231.2 | XP_006723294.1 | 4 - 5 | 647 | 132 | |
XM_011526997.1 | XP_011525299.1 | 4 - 5 | 647 | 132 | |
XM_011526999.1 | XP_011525301.1 | 4 - 5 | 647 | 132 | |
XM_011527001.1 | XP_011525303.1 | 4 - 5 | 647 | 132 | |
XM_011527002.1 | XP_011525304.1 | 4 - 5 | 647 | 132 | |
XM_011527006.1 | XP_011525308.1 | 4 - 5 | 647 | 132 | |
XM_011527007.1 | XP_011525309.1 | 4 - 5 | 647 | 132 | |
XM_017026841.1 | XP_016882330.1 | 4 - 5 | 647 | 132 | |
XM_017026842.1 | XP_016882331.1 | 4 - 5 | 647 | 132 | |
XM_017026843.1 | XP_016882332.1 | 4 - 5 | 647 | 132 | |
XM_017026844.1 | XP_016882333.1 | 1 - 2 | 423 | 132 | |
GenBank mRNA | AB054002.1 | - | 3 - 4 | 588 | 132 |
BC039016.1 | - | 4 - 5 | 623 | 132 | |
U77968.1 | - | 3 - 4 | 451 | 132 |