Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPAS1
Entrez Gene ID
4861
Gene Name
neuronal PAS domain protein 1
Gene Aliases
MOP5, PASD5, bHLHe11
Chromosome Location
Chr.19: 47019820 - 47045775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321086.1 NP_001308015.1 3 - 4 309 79
NM_002517.3 NP_002508.2 7 - 8 997 79
XM_006723231.2 XP_006723294.1 7 - 8 1021 79
XM_017026842.1 XP_016882331.1 7 - 8 1021 79
GenBank mRNA AB054002.1 - 6 - 7 962 79
AK299128.1 - 3 - 4 309 79
BC039016.1 - 7 - 8 997 79
U51628.1 - 4 - 5 475 79
U77968.1 - 6 - 7 825 79

View Full Product Details