The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003743.4 | NP_003734.3 | 3 - 4 | 348 | 55 |
NM_147223.2 | NP_671756.1 | 3 - 4 | 348 | 55 | |
NM_147233.2 | NP_671766.1 | 3 - 4 | 348 | 55 | |
XM_005264625.1 | XP_005264682.1 | 5 - 6 | 624 | 55 | |
XM_005264626.1 | XP_005264683.1 | 4 - 5 | 378 | 55 | |
XM_005264628.1 | XP_005264685.1 | 4 - 5 | 378 | 55 | |
XM_017005168.1 | XP_016860657.1 | 5 - 6 | 624 | 55 | |
XM_017005169.1 | XP_016860658.1 | 5 - 6 | 624 | 55 | |
GenBank mRNA | AJ000881.1 | - | 3 - 4 | 291 | 55 |
AJ000882.1 | - | 3 - 4 | 291 | 55 | |
AK290019.1 | - | 4 - 5 | 378 | 55 | |
BC111533.1 | - | 2 - 3 | 185 | 55 | |
BC111534.1 | - | 2 - 3 | 185 | 55 | |
U59302.1 | - | 3 - 4 | 284 | 55 | |
U90661.1 | - | 1 - 2 | 132 | 55 |