Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr.10: 7703273 - 7749520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002216.2 NP_002207.2 1 - 2 242 134
GenBank mRNA AK225277.1 - 1 - 2 105 134
AK225329.1 - 1 - 2 147 134
AK291379.1 - 1 - 2 190 134
BC132685.1 - 1 - 2 129 134
BG617651.1 - 1 - 2 243 134
X07173.1 - 1 - 2 143 134

View Full Product Details