The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002216.2 | NP_002207.2 | 20 - 21 | 2855 | 85 |
GenBank mRNA | AK225277.1 | - | 22 - 23 | 2718 | 85 |
AK225329.1 | - | 20 - 21 | 2760 | 85 | |
AK291379.1 | - | 20 - 21 | 2803 | 85 | |
BC132685.1 | - | 20 - 21 | 2742 | 85 | |
X07173.1 | - | 20 - 21 | 2756 | 85 |