Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr.10: 7703273 - 7749520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002216.2 NP_002207.2 8 - 9 1029 82
GenBank mRNA AK225277.1 - 10 - 11 892 82
AK225329.1 - 8 - 9 934 82
AK291379.1 - 8 - 9 977 82
AK297334.1 - 3 - 4 220 82
BC132685.1 - 8 - 9 916 82
X07173.1 - 8 - 9 930 82

View Full Product Details