The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004535.2 | NP_004526.1 | 11 - 12 | 2204 | 81 |
GenBank mRNA | AB020642.1 | - | 11 - 12 | 2204 | 81 |
BC062313.1 | - | 10 - 11 | 1164 | 81 |