Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYT1
Entrez Gene ID
4661
Gene Name
myelin transcription factor 1
Gene Aliases
C20orf36, MTF1, MYTI, NZF2, PLPB1, ZC2H2C1, ZC2HC4A
Chromosome Location
Chr.20: 64164474 - 64242253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004535.2 NP_004526.1 3 - 4 421 54
GenBank mRNA AB020642.1 - 3 - 4 421 54
BC062313.1 - 3 - 4 275 54

View Full Product Details