The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004535.2 | NP_004526.1 | 5 - 6 | 519 | 101 |
GenBank mRNA | AB020642.1 | - | 5 - 6 | 519 | 101 |
BC062313.1 | - | 5 - 6 | 373 | 101 |