The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277140.1 | NP_001264069.1 | 9 - 10 | 1086 | 135 |
NM_005721.4 | NP_005712.1 | 9 - 10 | 1295 | 135 | |
NR_102318.1 | 8 - 9 | 1170 | 135 | ||
GenBank mRNA | AB209353.1 | - | 7 - 8 | 804 | 135 |
AF006083.1 | - | 9 - 10 | 1121 | 135 | |
AF127773.1 | - | 9 - 10 | 1164 | 135 | |
AK300030.1 | - | 8 - 9 | 994 | 135 | |
AK300224.1 | - | 8 - 9 | 916 | 135 | |
AK302152.1 | - | 9 - 10 | 1040 | 135 | |
AK302359.1 | - | 9 - 10 | 1074 | 135 | |
AK303988.1 | - | 6 - 7 | 610 | 135 | |
AK312659.1 | - | 9 - 10 | 1095 | 135 | |
AK316213.1 | - | 9 - 10 | 1117 | 135 | |
AK316396.1 | - | 9 - 10 | 1086 | 135 | |
BC044590.1 | - | 9 - 10 | 1269 | 135 |