Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location
Chr.X: 111680741 - 111764164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099922.2 NP_001093392.1 18 - 19 2348 135
NM_001257230.1 NP_001244159.1 18 - 19 2244 135
NM_001257231.1 NP_001244160.1 18 - 19 2362 135
NM_001257234.1 NP_001244163.1 18 - 19 2317 135
NM_001257237.1 NP_001244166.1 18 - 19 2421 135
NM_001324292.1 NP_001311221.1 18 - 19 2348 135
NM_001324293.1 NP_001311222.1 16 - 17 2247 135
XM_006724693.2 XP_006724756.1 18 - 19 2721 135
XM_006724697.3 XP_006724760.2 18 - 19 2621 135
XM_006724698.3 XP_006724761.2 18 - 19 2746 135
XM_011531028.2 XP_011529330.1 18 - 19 2722 135
XM_011531030.1 XP_011529332.1 18 - 19 2720 135
XM_011531031.1 XP_011529333.1 18 - 19 2421 135
XM_011531032.1 XP_011529334.1 18 - 19 2227 135
XM_011531033.2 XP_011529335.1 18 - 19 2267 135
XM_011531034.2 XP_011529336.1 16 - 17 1961 135
XM_011531037.1 XP_011529339.1 8 - 9 978 135
XM_017029846.1 XP_016885335.1 18 - 19 2699 135
XM_017029847.1 XP_016885336.1 18 - 19 2722 135
XM_017029848.1 XP_016885337.1 18 - 19 2696 135
XM_017029849.1 XP_016885338.1 18 - 19 2721 135
XM_017029850.1 XP_016885339.1 18 - 19 2720 135
XM_017029851.1 XP_016885340.1 19 - 20 2629 135
XM_017029852.1 XP_016885341.1 11 - 12 1294 135
XM_017029853.1 XP_016885342.1 14 - 15 1536 135
XM_017029854.1 XP_016885343.1 5 - 6 1058 135
XM_017029855.1 XP_016885344.1 11 - 12 1986 135
GenBank mRNA AK223154.1 - 3 - 4 477 135
AK300394.1 - 18 - 19 2186 135
AK302729.1 - 18 - 19 2289 135
AK304712.1 - 18 - 19 2238 135
AK316306.1 - 18 - 19 2290 135
AK316522.1 - 18 - 19 2365 135
BC117377.1 - 3 - 4 475 135
BC117379.1 - 3 - 4 475 135

View Full Product Details