The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002738.6 | NP_002729.2 | 12 - 13 | 1598 | 79 |
NM_212535.2 | NP_997700.1 | 12 - 13 | 1598 | 79 | |
GenBank mRNA | BC036472.1 | - | 12 - 13 | 1558 | 79 |
M13975.1 | - | 12 - 13 | 1465 | 79 | |
X06318.1 | - | 12 - 13 | 1537 | 79 | |
X07109.1 | - | 12 - 13 | 1537 | 79 |