The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020820.3 | NP_065871.2 | 16 - 17 | 1904 | 99 |
XM_011528934.2 | XP_011527236.1 | 8 - 9 | 806 | 99 | |
GenBank mRNA | AB037836.2 | - | 16 - 17 | 1767 | 99 |
AJ320261.1 | - | 16 - 17 | 1904 | 99 | |
AL832913.1 | - | 9 - 10 | 873 | 99 | |
BC053616.1 | - | 12 - 13 | 1354 | 99 |