The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281737.1 | NP_001268666.1 | 10 - 11 | 1673 | 123 |
NM_001281738.1 | NP_001268667.1 | 9 - 10 | 1370 | 123 | |
NM_004453.3 | NP_004444.2 | 11 - 12 | 1814 | 123 | |
GenBank mRNA | AK293737.1 | - | 9 - 10 | 1355 | 123 |
AK304838.1 | - | 10 - 11 | 1435 | 123 | |
AK314803.1 | - | 11 - 12 | 1791 | 123 | |
BC011890.1 | - | 11 - 12 | 1616 | 123 | |
BX538129.1 | - | 12 - 13 | 1486 | 123 | |
EU093082.1 | - | 11 - 12 | 1510 | 123 | |
EU093083.1 | - | 3 - 4 | 428 | 123 | |
S69232.1 | - | 11 - 12 | 1578 | 123 |