The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281737.1 | NP_001268666.1 | 8 - 9 | 1326 | 119 |
NM_001281738.1 | NP_001268667.1 | 7 - 8 | 1023 | 119 | |
NM_004453.3 | NP_004444.2 | 9 - 10 | 1467 | 119 | |
GenBank mRNA | AK293737.1 | - | 7 - 8 | 1008 | 119 |
AK304838.1 | - | 8 - 9 | 1088 | 119 | |
AK314803.1 | - | 9 - 10 | 1444 | 119 | |
BC011890.1 | - | 9 - 10 | 1269 | 119 | |
BX538129.1 | - | 10 - 11 | 1139 | 119 | |
EU093082.1 | - | 9 - 10 | 1163 | 119 | |
S69232.1 | - | 9 - 10 | 1231 | 119 |