The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304351.1 | NP_001291280.1 | 1 - 2 | 153 | 69 |
NM_014265.5 | NP_055080.2 | 1 - 2 | 153 | 69 | |
NM_021777.4 | NP_068547.2 | 1 - 2 | 153 | 69 | |
NR_130709.1 | 1 - 2 | 153 | 69 | ||
NR_130710.1 | 1 - 2 | 153 | 69 | ||
XM_005273380.3 | XP_005273437.1 | 1 - 2 | 153 | 69 | |
XM_005273382.1 | XP_005273439.1 | 1 - 2 | 153 | 69 | |
XM_006716273.3 | XP_006716336.1 | 1 - 2 | 153 | 69 | |
XM_006716274.1 | XP_006716337.1 | 1 - 2 | 153 | 69 | |
XM_011544367.2 | XP_011542669.1 | 1 - 2 | 153 | 69 | |
XM_011544368.2 | XP_011542670.1 | 1 - 2 | 153 | 69 | |
XM_011544369.2 | XP_011542671.1 | 1 - 2 | 153 | 69 | |
XM_011544370.2 | XP_011542672.1 | 1 - 2 | 124 | 69 | |
XM_017012974.1 | XP_016868463.1 | 1 - 2 | 153 | 69 | |
XM_017012975.1 | XP_016868464.1 | 1 - 2 | 153 | 69 | |
XM_017012976.1 | XP_016868465.1 | 1 - 2 | 124 | 69 | |
GenBank mRNA | AF137334.1 | - | 1 - 2 | 82 | 69 |
AF137335.1 | - | 1 - 2 | 82 | 69 | |
AJ242015.1 | - | 1 - 2 | 90 | 69 | |
AK293382.1 | - | 1 - 2 | 124 | 69 | |
AK303437.1 | - | 1 - 2 | 124 | 69 | |
BC136478.1 | - | 1 - 2 | 69 | 69 | |
DA933009.1 | - | 1 - 2 | 154 | 69 |