The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001539.3 | NP_001530.1 | 2 - 3 | 316 | 134 |
GenBank mRNA | AK095541.1 | - | 7 - 8 | 134 | |
AK289623.1 | - | 2 - 3 | 244 | 134 | |
AY186741.1 | - | 2 - 3 | 242 | 134 | |
BC008182.1 | - | 2 - 3 | 229 | 134 | |
BI462555.1 | - | 2 - 3 | 311 | 134 | |
BM469197.1 | - | 2 - 3 | 217 | 134 | |
BT007292.1 | - | 1 - 2 | 125 | 134 | |
D13388.1 | - | 2 - 3 | 242 | 134 | |
DA003255.1 | - | 2 - 3 | 318 | 134 | |
L08069.1 | - | 2 - 3 | 207 | 134 |