The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001539.3 | NP_001530.1 | 3 - 4 | 502 | 64 |
GenBank mRNA | AJ277242.1 | - | 1 - 2 | 110 | 64 |
AK095541.1 | - | 6 - 7 | 64 | ||
AK289623.1 | - | 3 - 4 | 430 | 64 | |
AY186741.1 | - | 3 - 4 | 428 | 64 | |
BC008182.1 | - | 3 - 4 | 415 | 64 | |
BI462555.1 | - | 3 - 4 | 497 | 64 | |
BM469197.1 | - | 3 - 4 | 403 | 64 | |
BT007292.1 | - | 2 - 3 | 311 | 64 | |
D13388.1 | - | 3 - 4 | 428 | 64 | |
DA003255.1 | - | 3 - 4 | 504 | 64 | |
L08069.1 | - | 3 - 4 | 393 | 64 |