The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003400.3 | NP_003391.1 | 24 - 25 | 3800 | 135 |
XM_005264544.1 | XP_005264601.1 | 23 - 24 | 3777 | 135 | |
XM_005264546.1 | XP_005264603.1 | 22 - 23 | 2908 | 135 | |
XM_006712094.2 | XP_006712157.1 | 25 - 26 | 3943 | 135 | |
XM_011533097.1 | XP_011531399.1 | 24 - 25 | 3109 | 135 | |
XM_011533098.1 | XP_011531400.1 | 23 - 24 | 3642 | 135 | |
XM_011533099.2 | XP_011531401.1 | 24 - 25 | 4750 | 135 | |
GenBank mRNA | AK124769.1 | - | 13 - 14 | 2677 | 135 |
AK289920.1 | - | 24 - 25 | 3479 | 135 | |
AK299039.1 | - | 15 - 16 | 2202 | 135 | |
AK316481.1 | - | 15 - 16 | 2216 | 135 | |
AL833550.1 | - | 23 - 24 | 5745 | 135 | |
BC032847.2 | - | 25 - 26 | 3201 | 135 | |
BX647758.1 | - | 24 - 25 | 3782 | 135 | |
CR749840.1 | - | 25 - 26 | 3574 | 135 | |
D89729.1 | - | 23 - 24 | 3090 | 135 | |
Y08614.2 | - | 24 - 25 | 3110 | 135 |