Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL4
Entrez Gene ID
56062
Gene Name
kelch like family member 4
Gene Aliases
DKELCHL, KHL4
Chromosome Location
Chr.X: 87517541 - 87670050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057162.2 NP_476503.1 10 - 11 2278 130
GenBank mRNA AF284766.1 - 10 - 11 2185 130
BC140839.1 - 10 - 11 2147 130
BC140840.1 - 10 - 11 2147 130

View Full Product Details