The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 17 - 18 | 2215 | 75 |
NM_002209.2 | NP_002200.2 | 19 - 20 | 2467 | 75 | |
XM_005255313.1 | XP_005255370.1 | 19 - 20 | 2384 | 75 | |
XM_006721044.1 | XP_006721107.1 | 17 - 18 | 2169 | 75 | |
XM_011545849.1 | XP_011544151.1 | 19 - 20 | 2400 | 75 | |
GenBank mRNA | AK314246.1 | - | 19 - 20 | 2421 | 75 |
BC008777.2 | - | 17 - 18 | 2148 | 75 | |
Y00796.1 | - | 19 - 20 | 2379 | 75 |