The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 6 - 7 | 785 | 64 |
NM_002209.2 | NP_002200.2 | 8 - 9 | 1034 | 64 | |
XM_005255313.1 | XP_005255370.1 | 8 - 9 | 954 | 64 | |
XM_006721044.1 | XP_006721107.1 | 6 - 7 | 736 | 64 | |
XM_011545849.1 | XP_011544151.1 | 8 - 9 | 967 | 64 | |
GenBank mRNA | AK314246.1 | - | 8 - 9 | 988 | 64 |
BC008777.2 | - | 6 - 7 | 718 | 64 | |
Y00796.1 | - | 8 - 9 | 946 | 64 |