The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 7 - 8 | 931 | 77 |
NM_002209.2 | NP_002200.2 | 9 - 10 | 1180 | 77 | |
XM_005255313.1 | XP_005255370.1 | 9 - 10 | 1100 | 77 | |
XM_006721044.1 | XP_006721107.1 | 7 - 8 | 882 | 77 | |
XM_011545849.1 | XP_011544151.1 | 9 - 10 | 1113 | 77 | |
GenBank mRNA | AK314246.1 | - | 9 - 10 | 1134 | 77 |
BC008777.2 | - | 7 - 8 | 864 | 77 | |
Y00796.1 | - | 9 - 10 | 1092 | 77 |