Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
Chromosome Location
Chr.12: 6638076 - 6647414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032489.2 NP_115878.2 2 - 3 305 83
GenBank mRNA AB051833.1 - 2 - 3 305 83
AK302290.1 - 2 - 3 302 83
AK311572.1 - 2 - 3 302 83
AK312782.1 - 2 - 3 323 83
BC033010.2 - 2 - 3 302 83

View Full Product Details