Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDGFRP2
Entrez Gene ID
84717
Gene Name
hepatoma-derived growth factor-related protein 2
Gene Aliases
HDGF-2, HDGF2, HRP-2
Chromosome Location
Chr.19: 4472196 - 4502220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032631.3 NP_116020.1 15 - 16 2073 61
XM_011528377.2 XP_011526679.1 15 - 16 2433 61
GenBank mRNA BC000755.2 - 15 - 16 1963 61
BE745571.1 - - 341 61

View Full Product Details