The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139072.3 | NP_620711.3 | 2 - 3 | 731 | 77 |
XM_005246950.2 | XP_005247007.1 | 2 - 3 | 718 | 77 | |
GenBank mRNA | AF442487.1 | - | 2 - 3 | 644 | 77 |
AK289553.1 | - | 2 - 3 | 689 | 77 | |
AY358891.1 | - | 2 - 3 | 694 | 77 | |
BC035009.1 | - | 2 - 3 | 721 | 77 | |
DA400901.1 | - | 2 - 3 | 471 | 77 |